Order Entry
Czech Republic
ContactUsLinkComponent
Trypan blue 0.4% in PBS, dark blue solution
Trypan blue 0.4% in PBS, dark blue solution
Catalog # ICNA091691049
Supplier:  MP Biomedicals
CAS Number:  
undefined
Trypan blue 0.4% in PBS, dark blue solution
Catalog # ICNA091691049
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • pH:
    7.0-8.0
  • Osmolality:
    270-340mOsm
  • Cell Dye Uptake:
    ≥90%
  • Dye Concentration ((A594± 12nm):
    ≤0.5%

Specification Test Results

About this item

 :
 :960.82 g/mol
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969