Ghost Dye™ Red 710
Supplier: Tonbo Biosciences
Certificates
About this item
Ghost Dye Red 710 is an amine reactive viability dye, that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications.
This dye irreversibly binds free amines available on the cell surface, as well as intracellular free amines exposed in cells with compromised cell membranes.
Necrotic cells with compromised membranes will react with significantly more Ghost Dye Red 710 dye than viable cells in the same sample and therefore exhibit much greater fluorescence intensity, allowing exclusion of these cells from analysis.
Specifications
- Storage temperature:At ‒20 °C protected from light and moisture
- Application:Flow cytometry
Specifications
Frequently Bought Together