Order Entry
Czech Republic
ContactUsLinkComponent
PicoKine Mouse CD23/FCER2; ELISA Kit
PicoKine Mouse CD23/FCER2; ELISA Kit
Katalogové číslo BSBTEK0924
Dodavatel:  Boster Bio
Číslo CAS:  
undefined
PicoKine Mouse CD23/FCER2; ELISA Kit
Katalogové číslo BSBTEK0924
Dodavatel:  Boster Bio
Číslo CAS:  

Specifikace

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    CD23/FCER2
  • Description:
    Mouse CD23/FCER2 PicoKine; ELISA kit
  • Tests per kit:
    96 wells/kit

Specifikace

O tomto produktu

Sandwich high sensitivity ELISA kit for quantitative detection of mouse CD23/FCER2.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect mouse Fc Epsilon Rii/Fcer2 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Fc Epsilon Rii/Fcer2 tissue specificity:

96wells/kit, with removable strips.