Order Entry
Czech Republic
ContactUsLinkComponent
QuickDetect™ Acetylcholine (ACh) (Human) ELISA Kit
QuickDetect™ Acetylcholine (ACh) (Human) ELISA Kit
Catalog # ABCAAB287811-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ Acetylcholine (ACh) (Human) ELISA Kit
Catalog # ABCAAB287811-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Host:
  • Primary antibody reactivity:
    Human
  • Description:
    QuickDetect™ Acetylcholine (ACh) (Human) ELISA Kit, 100 assays
  • Environmentally Preferable:

Specifications

About this item

A rapid ELISA kit to quantify Acetylcholine in human serum, plasma, urine, cell culture samples.

  • Easy, convenient and time-saving method to measure the level of Acetylcholine in human Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 3 to 160 pg/ml
  • Sensitivity: <0,5 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed

Acetylcholine (ACh) is an organic chemical that functions in the brain and body of many types of animals, including humans, as a neurotransmitter-a chemical released by nerve cells to send signals to other cells. Acetylcholine is also used as a neurotransmitter in the autonomic nervous system, both as an internal transmitter for the sympathetic nervous system and as the final product released by the parasympathetic nervous system. In the brain, acetylcholine functions as a neurotransmitter and as a neuromodulator. The brain contains a number of cholinergic areas, each with distinct functions. They play an important role in arousal, attention, memory and motivation. BioVision’s Acetylcholine ELISA kit is a sandwich ELISA assay for the quantitative measurement of ACh in human serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Acetylcholine captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.