Order Entry
Czech Republic
ContactUsLinkComponent
Iodine concentrate for dilution to 0.05 mol (0.1N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Health
Iodine concentrate for dilution to 0.05 mol (0.1N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Katalogové číslo 91351.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Iodine concentrate for dilution to 0.05 mol (0.1N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Katalogové číslo 91351.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Molarity:
    0.04975 - 0.05025 mol/l (20°C)

Specification Test Results

About this item

 :
 :253.81 g/mol
 :Dose for preparation of 1 litre of solution.
 :GHS08
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355