ContactUsLinkComponent
Cleanroom Hoods, Flat Top
undefined
Cleanroom Hoods, Flat Top
  • Catalog No:
  • ALSO1003835-S
  • ALSO1003835-XS
  • ALSO1003835-L
  • ALSO1003835-M
  • ALSO1003835-XXL
  • ALSO1003835-XL
  • ALSO1002390-XS
  • ALSO1003835-3XL
  • ALSO1002390-M
  • ALSO1002390-S
  • ALSO1002390-XL
  • ALSO1002390-L
  • ALSO1002390-3XL
  • ALSO1002390-XXL
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Weight:
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
Frequently Bought Together