3'-Bromo-4'-(trifluoromethyl)acetophenone ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1807875-04-2
:
Formula:C₉H₆BrF₃O
MDL Number:MFCD28154820
:267.04254 g/mol
Frequently Bought Together
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane, liquid, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
EX-CELL Advanced CHO Fed-batch Medium 1 * 1.000 mL
Supplier: Merck
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals