2,4-Dibromo-6-fluorobenzoic acid ≥97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:183065-69-2
:
Formula:C₇H₃Br₂FO₂
MDL Number:MFCD03094087
:297.90 g/mol
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals