Order Entry
Czech Republic
ContactUsLinkComponent
 
Water, NORMATOM®, Ultrapure for trace metal analysis
Catalog # 83880.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water, NORMATOM®, Ultrapure for trace metal analysis
Catalog # 83880.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Colouration (APHA):
    max. 10
  • Cl (Chloride):
    max. 1 ppb
  • PO4 (Phosphate):
    max. 1 ppb
  • SO4 (Sulphate):
    max. 1 ppb
  • Ag (Silver):
    max. 10 ppt
  • Al (Aluminium):
    max. 20 ppt
  • As (Arsenic):
    max. 10 ppt
  • Au (Gold):
    max. 10 ppt
  • B (Boron):
    max. 20 ppt
  • Ba (Barium):
    max. 10 ppt
  • Be (Beryllium):
    max. 10 ppt
  • Bi (Bismuth):
    max. 10 ppt
  • Ca (Calcium):
    max. 10 ppt
  • Cd (Cadmium):
    max. 10 ppt
  • Ce (Cerium):
    max. 10 ppt
  • Co (Cobalt):
    max. 10 ppt
  • Cr (Chromium):
    max. 10 ppt
  • Cs (Cesium):
    max. 10 ppt
  • Cu (Copper):
    max. 10 ppt
  • Dy (Dysprosium):
    max. 1 ppt
  • Er (erbium):
    max. 1 ppt
  • Eu (Europium):
    max. 1 ppt
  • Fe (Iron):
    max. 10 ppt
  • Ga (Gallium):
    max. 10 ppt
  • Gd (Gadolinium):
    max. 1 ppt
  • Ge (Germanium):
    max. 10 ppt
  • Hf (Hafnium):
    max. 1 ppt
  • Hg (Mercury):
    max. 20 ppt
  • Ho (Holmium):
    max. 1 ppt
  • In (Indium):
    max. 1 ppt
  • K (Potassium):
    max. 10 ppt
  • La (Lanthanum):
    max. 1 ppt
  • Li (Lithium):
    max. 10 ppt
  • Lu (Lutetium):
    max. 1 ppt
  • Mg (Magnesium):
    max. 10 ppt
  • Mn (Manganese):
    Max. 20 ppt
  • Mo (Molybdenum):
    max. 10 ppt
  • Na (Sodium):
    max. 10 ppt
  • Nb (Niobium):
    max. 10 ppt
  • Nd (Neodymium):
    max. 1 ppt
  • Ni (Nickel):
    max. 10 ppt
  • Pb (Lead):
    max. 10 ppt
  • Pd (Palladium):
    max. 10 ppt
  • Pr (Praseodymium):
    max. 10 ppt
  • Pt (Platinum):
    max. 10 ppt
  • Rb (Rubidium):
    max. 10 ppt
  • Re (Rhenium):
    max. 10 ppt
  • Rh (Rhodium):
    max. 10 ppt
  • Ru (Ruthenium):
    max. 10 ppt
  • Sb (Antimony):
    max. 10 ppt
  • Sc (Scandium):
    max. 10 ppt
  • Se (Selenium):
    max. 50 ppt
  • Sm (Samarium):
    max. 10 ppt
  • Sn (Tin):
    max. 10 ppt
  • Sr (Strontium):
    max. 10 ppt
  • Ta (Tantalum):
    max. 10 ppt
  • Tb (Terbium):
    max. 10 ppt
  • Te (Tellurium):
    max. 1 ppt
  • Th (Thorium):
    max. 1 ppt
  • Ti (Titanium):
    max. 10 ppt
  • Tl (Thallium):
    max. 10 ppt
  • Tm (Thulium):
    max. 10 ppt
  • U (Uranium):
    max. 1 ppt
  • V (Vanadium):
    max. 10 ppt
  • W (Tungsten):
    max. 10 ppt
  • Y (Yttrium):
    max. 1 ppt
  • Yb (Ytterbium):
    max. 10 ppt
  • Zn (Zinc):
    max. 10 ppt
  • Zr (Zirconium):
    max. 10 ppt

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332