Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Tube rollers
VWR®, Tube rollers
Supplier:  VWR Collection
Certificates
undefined
VWR®, Tube rollers
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 444-1339
  • 444-1342
  • Speed:
  • 2 - 80 min⁻¹ (Variable)
  • 2 - 80 min⁻¹ (Variable)
  • Dimensions:
  • Individual roller: 30×320 mm
  • Individual roller: 30×320 mm
  • W×D×H:
  • 457×178×89 mm
  • 457×356×89 mm
  • Operating temperature:
  • 4...40 °C
  • 4...40 °C
  • Load capacity:
  • 2 kg
  • 4 kg

Specifications

Frequently Bought Together