Order Entry
Czech Republic
ContactUsLinkComponent
Water ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Water ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Catalog # 92499.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Catalog # 92499.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Resistivity (at filling; @ 25°C):
    min. 18 Mohm
  • TOC (at filling):
    max. 10 ppb
  • pH (at filling; @25°C):
    5.5 - 8.0
  • Evaporation residue:
    max. 0.00005%
  • Gradient use test (205 nm):
    max. 0.002 AU
  • Gradient use test (254 nm):
    max. 0.0005 AU
  • Baseline drift (205 nm):
    max. 0.02 AU
  • Ag (Silver):
    max. 10 ppb
  • Al (Aluminium):
    max. 25 ppb
  • Ca (Calcium):
    max. 25 ppb
  • Cu (Copper):
    max. 10 ppb
  • Fe (Iron):
    max. 10 ppb
  • K (Potassium):
    max. 10 ppb
  • Mg (Magnesium):
    max. 10 ppb
  • Mn (Manganese):
    max. 10 ppb
  • Na (Sodium):
    max. 50 ppb
  • Ni (Nickel):
    max. 10 ppb
  • Pb (Lead):
    max. 10 ppb
  • Zn (Zinc):
    max. 10 ppb
  • Suitability for PFAS analysis:
    passes test
  • Suitability for LC-MS:
    passes test

Specification Test Results

About this item

CAS Number:7732-18-5
 :
Formula:H₂O
MDL Number:MFCD00011332
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
Storage Temperature:Ambient
EINECS:231-791-2