Crotamiton for system suitability CRS, EDQM
EDQM0002338
: EDQM
:
- Pk:1 mg
- CAS No.:483-63-6
- Description:Crotamiton for system suitability CRS
- Storage temperature:-20 °C+/-5 °C
Reference Standard for laboratory tests
Reference Standard for laboratory tests