Order Entry
Czech Republic
ContactUsLinkComponent
 
Elastic adjustable cam-lock wrist band
  115-3039
 :  DESCO
Elastic adjustable cam-lock wrist band
  115-3039
 :  DESCO

 

  • Colour:
    Blue
  • Description:
    Elastic adjustable cam-lock wrist band, 10 mm
  • Pk:
    1

 

 

  • Versatile; metal back or plastic back
  • Metal helps contact to dry skin - insert plastic cover if there are metal-on-skin issues
  • Adjustable band to suit wrist circumference of 130 to 180 mm
  • 10 mm stud connector only