ContactUsLinkComponent
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Certificates
undefined
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Certificates
Specifications
  • Connection type (port 1):
  • Male luer lock
  • Male luer lock
  • Connection type (port 2):
  • ¹/₄-28 UNF
  • ¹/₄-18 NPT thread
  • No. of ports:
  • 2
  • 2