Value Plastics® Adapter Fittings, Female Luer to Threaded, Straight
Easily transition between luer fittings and UNF or NPT threads.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
- Environmentally Preferable:
- Catalog No:
- MFLX30800-00BLK
- MFLX45508-76BLK
- Connection type (port 1):
- Female luer
- Female luer
- Connection type (port 2):
- 1/8-27-NPT thread
- 1/4-28 UNF bottom sealing thread
- No. of ports:
- 2
- 2
- Warranty:
- 3 Months
Frequently Bought Together