ContactUsLinkComponent
Stainless steel probe
Stainless steel probe
Catalog # 115-3063
Supplier:  DESCO
Stainless steel probe
Catalog # 115-3063
Supplier:  DESCO
Specifications
  • Description:
    Stainless steel probe, hooked
  • Pk:
    1
Specifications
About this item
Hooked stainless steel probe, 140 mm.
  • Ergonomic hex handle
  • Allows operator to better grip probe; reduces fatigue from gripping, product won't roll
  • Non-magnetic, conductive, 300 series stainless steel
  • Stainless steel won’t corrode - no problem in working with magnetised parts