Order Entry
Czech Republic
ContactUsLinkComponent
Humidity indicator 3 spot cards
Humidity indicator 3 spot cards
Catalog # 129-2643
Supplier:  DESCO
Humidity indicator 3 spot cards
Catalog # 129-2643
Supplier:  DESCO

Specifications

  • Description:
    Humidity indicator 3 spot cards indicates: 10, 50, 60%
  • Pk:
    125

Specifications

About this item

Humidity indicator cards (HICs) are printed with moisture sensitive spots which respond to various levels of humidity with a visible colour change from blue to pink.

  • The moisture sensitive spots are made of blotting paper impregnated with cobaltous chloride
  • The humidity inside dry packed barrier bags can be monitored by including an HIC inside

When the bag is opened, the card can be examined for proper dryness inside the bag.

L×W: 76,2×50,8 mm (3×2 ").

Certifications: Complies with IPC/ JEDEC J-STD-033D.