VWR®, Endosulfan II, Certified Reference Material (Neat)
92471.0010
:
- Pack type:Glass ampoule/vial
- Pk:10 mg
- Standards:CRM ISO 17034
- CAS No.:33213-65-9
- Description:Endosulfan II, Certified Reference Material (Neat)
- Storage conditions:Ambient
- Hazard class description:Toxic
- Shipping conditions:Ambient temperature
- MDL No.:MFCD00151176
- UN No.:2811
- Formula:C9H6Cl6O3S
- Molecular weight:406.91 g/mol
- Description:Endosulfan II
: Certified Reference Material ISO 17034