


Trifluoroborane acetonitrile complex, Sigma-Aldrich®
Catalog # 8.14784.0100
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Assay BF3 (acidimetric):15.0 - 19.0 %
- Density (d 20 °C/ 4 °C):0.870 - 0.880
Specification Test Results
About this item
:
:108.858 g/mol
Boiling Pt:82 °C (1013 hPa)
Density:0.875 g/cm3 (20 °C)
ADR:3 (6.1, 8),II
:M-Clarity™ quality level = MQ200
Flash Pt:10 °C
UN:3286
:GHS06
:GHS05
:GHS02
CAS Number:420-16-6
Formula:BF₃·CH₃CN
MDL Number:MFCD28125563