Order Entry
Czech Republic
ContactUsLinkComponent
 
Recombinant COL11A1 (from E. coli)
Catalog # NOVUNBP2-58159PEP
Supplier:  Novus Biologicals
CAS Number:  
Recombinant COL11A1 (from E. coli)
Catalog # NOVUNBP2-58159PEP
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Amino acid number:
    DCDSSAPKAAQAQEPQIDEYAPEDIIEYDYEYGEAEYKEAESVTEGPTVTEETIAQTEANIVDDFQEYNYG
  • Protein synonyms:
    COLL6|collagen, type XI, alpha 1|alpha-1 polypeptide
  • Protein/peptide name:
    COL11A1
  • Purity:
    >80% by SDS-PAGE and Coomassie blue staining
  • Formulation:
    PBS and 1M Urea, pH 7.4. No Preservative

Specifications

About this item

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human COL11A1.

The COL11A1 gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. Type XI collagen is aheterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Mutations in thisgene are associated with