2,4,4'-Trichlorobiphenyl (PCB No. 28), Sigma-Aldrich®
Catalog # SIALBCR291-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
ADR:9,II
:257.55 g/mol
Storage Temperature:Refrigerator
EINECS:230-293-2
UN:2315
CAS Number:7012-37-5
Formula:C₁₂H₇Cl₃