2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153), Sigma-Aldrich®
Catalog # SIALBCR297-25MG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 mg
- Sekvence:2315
Specifications
About this item
:
Boiling Pt:259 °C (1013 hPa)
Melting Pt:103…104 °C
Storage Temperature:Refrigerator
UN:3432
ADR:9,II
CAS Number:35065-27-1
Formula:C₁₂H₄Cl₆