

2-(2-Aminoethyl)-1,3-dioxolane ≥98.0% (by GC, titration analysis)
Catalog # ABCRTCA1176.0005
Supplier: abcr
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:5 g
- Sekvence:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:75°C
Storage Temperature:Refrigerator
EINECS:227-277-2
UN:2733
ADR:3,III
:GHS02
:GHS05
CAS Number:5754-35-8
Formula:C₅H₁₁NO₂
MDL Number:MFCD00142756