ContactUsLinkComponent
Propagyl bromide ≥80% in toluene stabilised
  ABCRAB110394.0050
 :  abcr
 :  
Propagyl bromide ≥80% in toluene stabilised
  ABCRAB110394.0050
 :  abcr
 :  
  • Pack type:
    Glass bottle
  • Pk:
    50 mL
 :
 :118.96 g/mol
 :88…90 °C
 :–61 °C
 :1.335
 :3 °C (37 °F)
 :Refrigerator
 :203-447-1
 :2924
 :3,II
 :Stabiised with magnesium oxide (MgO)
 :GHS02
 :GHS06
 :GHS08
 :GHS05
 :GHS07
 :106-96-7
 :C₃H₃Br
 :MFCD00000241