Order Entry
Czech Republic
ContactUsLinkComponent
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
Toxic
Health
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
Catalog # ABCR22773.0050
Supplier:  abcr
CAS Number:  
Toxic
Health
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
Catalog # ABCR22773.0050
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

About this item

 :
 :202.55 g/mol
Boiling Pt:14…16 °C (4 mmHg)
Melting Pt:∼20…25 °C
Density:1.687
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:210-223-7
UN:1577
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:610-40-2
Formula:C₆H₃ClN₂O₄
MDL Number:MFCD00007212