Order Entry
Czech Republic
ContactUsLinkComponent
cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # ABCRTCA1448.0005
Supplier:  abcr
CAS Number:  
cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # ABCRTCA1448.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :143.19 g/mol
CAS Number:3685-23-2
Formula:C₇H₁₃NO₂
MDL Number:MFCD00191730