Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Acetone

Acetone

Glass bottle, 10 L
Catalog # MACH20001CT0M50001
Flammable
Irritant
Supplier:   MACH
Acetone
Catalog # MACH20001CT0M50001
Supplier:  MACH
CAS Number:  67-64-1
Acetone
Acetone

Glass bottle, 10 L

CAS Number:  67-64-1
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.