Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Barium nitrate

Barium nitrate

Plastic bottle for solids, 10 g
Catalog # MACH30026AP0G0010
Oxidizing
Irritant
Supplier:   MACH
Barium nitrate
Catalog # MACH30026AP0G0010
Supplier:  MACH
CAS Number:  10022-31-8
Barium nitrate

Plastic bottle for solids, 10 g

CAS Number:  10022-31-8
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:10022-31-8
  • Formula:Ba(NO₃)₂
  • MDL Number:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:13,00986
  • Melting Pt:∼592 °C
  • Density:∼3.23 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-020-5
  • UN:1446
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.