Ammonium iron(III) sulphate dodecahydrate
Catalog # MACH30163AP0G0500
Supplier: MACH
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:482.2 g/mol
Merck Index:12,00549
Boiling Pt:230 °C (1013 hPa)
Melting Pt:39…41 °C
Density:1.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-382-4
:GHS07
CAS Number:7783-83-7
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004