Calcium sulphate dihydrate
Catalog # MACH30190AP0G1000
Supplier: MACH
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:172.17 g/mol
Merck Index:13,01711
Density:2.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-900-3
CAS Number:10101-41-4
Formula:CaSO₄·2H₂O
MDL Number:MFCD00010912