0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
ABCRAB109899.0002
:
abcr
:
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
ABCRAB109899.0002
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
2 g
:
:
671.70 g/mol
:
190 °C (dec.)
:
235-170-7
:
12112-67-3
:
C₁₆H₂₄Cl₂Ir₂
:
MFCD00012414