Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, 2, 2', 3, 3',4,4', 5, 5'-Octabromobiphenyl, 35 µg/ml in isooctane, CRM
VWR®, 2, 2', 3, 3',4,4', 5, 5'-Octabromobiphenyl, 35 µg/ml in isooctane, CRM
Catalog # 92420.010
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, 2, 2', 3, 3',4,4', 5, 5'-Octabromobiphenyl, 35 µg/ml in isooctane, CRM
Catalog # 92420.010
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule/vial
  • Pk:
    1 mL
  • Standards:
    CRM ISO 17034
  • CAS No.:
    67889-00-3
  • Description:
    2, 2', 3, 3',4,4', 5, 5'-Octabromobiphenyl, 35 µg/ml in isooctane
  • Storage conditions:
    Ambient (>5 °C)
  • UN No.:
    1262
  • Description:
    2,2',3,3',4,4',5,5'-Octabromobiphenyl

Specifications

About this item

Certifications: Certified reference material.
Caution: Highly Flammable.