VWR®, Saytex BT-93, 50 µg/ml in toluene, CRM
Catalog # 92140.010
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule/vial
- Pk:1 mL
- Standards:CRM ISO 17034
- CAS No.:32588-76-4
- Description:Saytex BT-93, 50 µg/ml in toluene
- Storage conditions:Ambient (>5 °C)
- UN No.:1294
- Description:Saytex BT-93
Specifications
About this item
Certifications: Certified reference material.
Caution: Highly Flammable.