VWR®, FireMaster 680, 100 µg/ml in toluene, CRM
92111.010
:
- Pack type:Glass ampoule/vial
- Pk:1 mL
- Standards:CRM ISO 17034
- CAS No.:37853-59-1
- Description:FireMaster 680, 100 µg/ml in toluene
- Storage conditions:Ambient (>5 °C)
- UN No.:1294
- Description:FireMaster 680
: Certified reference material.
: Highly Flammable.