Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Ethyl oleate (neat)
VWR®, Ethyl oleate (neat)
Catalog # 91649.0100
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Ethyl oleate (neat)
Catalog # 91649.0100
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule/vial
  • Pk:
    100 mg
  • CAS No.:
    111-62-6
  • Description:
    Ethyl oleate (neat)
  • Storage conditions:
    Refrig (0 - 5 °C)
  • Formula:
    C20H38O2
  • Molecular weight:
    310,51
  • Description:
    Ethyl oleate

Specifications

About this item