ContactUsLinkComponent
Welding sweatbands
Welding sweatbands
Catalog # MMMAX5-SWBD01
Supplier:  3M
undefined
Welding sweatbands
Catalog # MMMAX5-SWBD01
Supplier:  3M
Specifications
  • For:
    3M™ X5000/X5500 safety helmets
  • Description:
    Nylon welding sweatbands
  • Pk:
    1
Specifications
About this item
This replacement sweatband fits the 3M™ SecureFit™ safety helmet models X5000 and X5500.
  • Reversible replacement sweatband
  • Can be removed for cleaning
  • Fits 3M SecureFit™ safety helmet series X5000 and X5500
The sweatband is reversable and removable for cleaning or replacement. Original equipment replacement part for the 3M™ SecureFit™ safety helmet models X5000 and X5500.