ContactUsLinkComponent
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Horizontal
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Horizontal
Catalog # 99901.010
Supplier:  VWR Chemicals
undefined
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Horizontal
Catalog # 99901.010
Supplier:  VWR Chemicals
Specifications
  • Description:
    Withdrawal system for stainless steel barrels 10 L (including dip tube)
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 L stainless steel barrels.
  • Withdrawal system for 10 L barrels (stainless steel)
  • Perfluorelastomer O-rings
  • Includes 1 dip tube for 10 L barrels
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 10 L returnable stainless steel barrels.