

4-Chlorobiphenyl (PCB No. 3) ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2051-62-9
:
Formula:C₁₂H₉Cl
MDL Number:MFCD00013640
Boiling Pt:282 °C
Melting Pt:76…77 °C
:188.66 g/mol
EINECS:218-127-7
:GHS07
:GHS09
Frequently Bought Together