ContactUsLinkComponent
Cleanroom hoods, sterile, CleanMax®
Cleanroom hoods, sterile, CleanMax®
Catalog # 114-9542
undefined
Cleanroom hoods, sterile, CleanMax®
Catalog # 114-9542
Specifications
  • Clothing Size:
    One size
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Description:
    Clean sterile hood
  • Seq:
    02
  • Pk:
    100 PAIR
  • Cleanroom class:
    100/ISO 5
Specifications
About this item
Clean and Sterile disposable hoods are individually double-bagged and folded to promote easier gowning with less excursions.
  • Clean and sterile in silicone-free environment
  • Latex-free
  • Covers shoulders
  • One size
  • Ties to customize fit
  • Individually double-bagged
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.