ContactUsLinkComponent
Cleanroom sleeves, CleanMax®
Cleanroom sleeves, CleanMax®
Catalog # 114-9544
undefined
Cleanroom sleeves, CleanMax®
Catalog # 114-9544
Specifications
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Length:
    18"
  • Pk:
    50 PAIR
  • Cleanroom class:
    100/ISO 5
Specifications
About this item
Clean manufactured nonsterile sleeves with bound seams. Clean manufactured in a silicone-free environment.
  • Fluid repellent; tested in accordance with ASTM D6978 for chemotherapy permeation
  • Open fold for easier donning
  • Resistant to blood, body fluid and viral penetration
  • Resistant to blood borne pathogens
  • IEST-RP-CC003 Category I particle cleanliness
  • Latex-free and silicone-free
CleanMax® cleanroom garments for controlled environments provide optimal comfort, quality, and protection. Ideally suited for use in ISO Class 4 - 8 cleanrooms and all controlled environments. CleanMax® garments are easy to don, comfortable to wear, and are backed by Lakeland's 30+ years as a manufacturer of disposable protective apparel.
Packaging: Packaged in pairs in a protective outer bag for ante areas.