Order Entry
Czech Republic
ContactUsLinkComponent
Magbeads FastDNA kit, 192 preparations, MP Biomedicals
Magbeads FastDNA kit, 192 preparations, MP Biomedicals
Catalog # 116575192.
Supplier:  MP Biomedicals
CAS Number:  
undefined
Magbeads FastDNA kit, 192 preparations, MP Biomedicals
Catalog # 116575192.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    192 Tests
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Magbeads FastDNA kit

Specifications

About this item

MagBeads FastDNA® Kit is intended for rapid extraction of DNA from tissue, cells, blood, saliva, swabs, blood spots, semen, and other clinical samples. DNA can be used directly for PCR, quantitative PCR, Southern Blot, detection of viral DNA and so on.

  • High quality DNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals

MagBeads FastDNA® Kit is based on the purification method of high binding magnetic particles. The sample is lysed and digested. RNA is released into the lysate. After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.

The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts. Finally the RNA was eluted with elution buffer.