Order Entry
Czech Republic
ContactUsLinkComponent
Zirconyl chloride octahydrate 98%, Reagent Grade, Sigma-Aldrich®
Corrosive
Zirconyl chloride octahydrate 98%, Reagent Grade, Sigma-Aldrich®
Katalogové číslo SIAL224316-5G
Supplier:  Merck
CAS Number:  
Corrosive
Zirconyl chloride octahydrate 98%, Reagent Grade, Sigma-Aldrich®
Katalogové číslo SIAL224316-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :322.25 g/mol
Melting Pt:> 150 °C
Density:1.91 g/cm³ (20 °C)
EINECS:231-717-9
UN:3260
ADR:8,III
 :GHS05
CAS Number:13520-92-8
Formula:Cl₂OZr·8H₂O
MDL Number:MFCD00149898