Orange G, certified by the Biological Stain Commission
Catalog # SIAL861286-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:452.38 g/mol
Melting Pt:141 °C
Density:0.45…0.47 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-705-6
CAS Number:1936-15-8
Formula:C₁₆H₁₀N₂Na₂O₇S₂
MDL Number:MFCD00012457