Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Glutamine ≥98.5% FCC
L(+)-Glutamine ≥98.5% FCC
Catalog # SPCMG1039-1KG
Supplier:  Spectrum Chemical
CAS Number:  
L(+)-Glutamine ≥98.5% FCC
Catalog # SPCMG1039-1KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specification Test Results

About this item

 :
 :146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044