Order Entry
Czech Republic
ContactUsLinkComponent
 
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.1% in aqueous solution indicator
Catalog # SIAL318922-100ML
Supplier:  Merck
CAS Number:  
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.1% in aqueous solution indicator
Catalog # SIAL318922-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :692.54 g/mol
EINECS:238-676-6
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428