D-Glucono-1,5-lactone ≥99.0% USP
Catalog # SPCMG1165-1KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 101.0%
- Lead (Pb):max. 0.001%
- Heavy Metals:max. 0.002%
- Reducing Substances:To Pass Test
- Identification:To Pass Test
Specification Test Results
About this item
Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
:
:178.14 g/mol
Storage Temperature:Ambient
CAS Number:90-80-2
Formula:C₆H₁₀O₆
MDL Number:MFCD00006647
Boiling Pt:385
Melting Pt:153
Merck Index:13,04470