Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium chloride ≥99.0%, granular FCC
Irritant
Ammonium chloride ≥99.0%, granular FCC
Catalog # SPCMA1167-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Ammonium chloride ≥99.0%, granular FCC
Catalog # SPCMA1167-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay (Dried Basis):
    99.0 % min.
  • Lead (Pb):
    max.4 mg/kg
  • Loss On Drying:
    max. 0.5 %
  • Identification:
    To Pass Test

Specification Test Results

About this item

 :
 :53.49 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00143604