

Ethyl acetate ≥99.9% for HPLC
Catalog # SIAL650528-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle, narrow mouth
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
About this item
:
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
:GHS02
:GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171