Diammonium hydrogen orthophosphate ≥96.0% FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay [(NH₄)₂HPO₄]:96.0 - 102.0 %
- Arsenic (As):max. 3 mg/kg
- Fluoride:max. 10 mg/kg
- Lead (Pb):max. 4 mg/kg
- Identification:To Pass Test
Specification Test Results
Frequently Bought Together