


Oxidation reagent 0.05 M I₂ in pyridine/water (90/10) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Boiling Pt:115.25 °C (1.013 hPa.) (main component)
Melting Pt:–42.2 °C (main component)
Density:0.983 g/cm3 (20 °C) (main component)
Flash Pt:12.4 %(V) (main component)
UN:1993
ADR:3,III
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together